Learn More
Cayman Chemical 3-AcetylpyrIdIn NAD 25mg
SDP
Catalog No. 502995660
Encompass_Preferred
A NAD analog; used to study the mechanisms of oxidative phosphorylation
Catalog No. 50-299-5660 Supplier Cayman Chemical Supplier No. 2188425 MG
A NAD analog; used to study the mechanisms of oxidative phosphorylation