Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylpyridine, 98%

Catalog No. AAA1424636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1424636 500 g
AAA1424614 25 g
AAA1424622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1424636 Supplier Thermo Scientific Chemicals Supplier No. A1424636
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 350-03-8 | C7H7NO | 121.139 g/mol

3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.

Solubility
Soluble in Chloroform. Miscible with water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006396
InChI Key WEGYGNROSJDEIW-UHFFFAOYSA-N
Synonym 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
PubChem CID 9589
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)C1=CN=CC=C1

Specifications

Melting Point 12°C to 13°C
Density 1.106
Boiling Point 218°C to 220°C
Flash Point 104°C (219°F)
Odor Characteristic
Refractive Index 1.535
Quantity 500 g
UN Number UN2810
Beilstein 107751
Merck Index 14,6116
Solubility Information Soluble in Chloroform. Miscible with water.
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material 3-Acetylpyridine
Show More Show Less
Hazard Category H300-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-496-7
RTECSNumber OB5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.