Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylthiazolidine-2-thione 98.0+%, TCI America™
SDP

Catalog No. A23015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23015G Supplier TCI America Supplier No. A23015G

Chemical Identifiers

CAS 76397-53-0
Molecular Formula C5H7NOS2
Molecular Weight (g/mol) 161.237
InChI Key BMOBFBQWISCVKX-UHFFFAOYSA-N
PubChem CID 11789521
IUPAC Name 1-(2-sulfanylidene-1,3-thiazolidin-3-yl)ethanone
SMILES CC(=O)N1CCSC1=S

Specifications

Quantity 5 g
Formula Weight 161.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Acetylthiazolidine-2-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.