Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Acetylthio)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. A31341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31341G 1 g
A31345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31341G Supplier TCI America Supplier No. A31341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41345-70-4
Molecular Formula C5H8O3S
Molecular Weight (g/mol) 148.176
MDL Number MFCD06208418
InChI Key AYQXANXXZYKTDL-UHFFFAOYSA-N
Synonym 3-(Acetylthio)propanoic Acid
PubChem CID 262719
IUPAC Name 3-acetylsulfanylpropanoic acid
SMILES CC(=O)SCCC(=O)O

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 128°C
Quantity 1 g
Formula Weight 148.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Acetylthio)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.