Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Acetylthiophene | 1468-83-3 | MFCD00005468 | 126.18 | 10 G
SDP

Catalog No. 5000460434
Encompass_Preferred

3-Acetylthiophene is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.

  • Solid appearance
  • Light brown to brown color
  • Chemical formula: C6H6OS
  • Soluble in DMSO
  • Store at 4°C, protected from light
  • For in-solvent storage, -80°C for 6 months or -20°C for 1 month
  • Ships at room temperature (continental US)

Catalog No. 50-004-60434 Supplier Medchemexpress LLC Supplier No. HYW00736110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Acetylthiophene is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.

  • Solid appearance
  • Light brown to brown color
  • Chemical formula: C6H6OS
  • Soluble in DMSO
  • Store at 4°C, protected from light
  • For in-solvent storage, -80°C for 6 months or -20°C for 1 month
  • Ships at room temperature (continental US)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.