Learn More
Cayman Chemical 3-AcetylursolIc AcId 10mg
SDP
Catalog No. 502988279
Encompass_Preferred
A triterpene with diverse biological activities; inhibits ADP-, thrombin-, or epinephrine-induced aggregation of isolated rat platelets (IC50s = P. falciparum in vitro (IC50 = 4 µM); active against B. cereus, S. aureus, and S. pneumoniae (MICs = 39.1, 19.6, and 4.81 µM, respectively); cytotoxic to HepG2 cells (LC50 = 351 µM)
Catalog No. 50-298-8279 Supplier Cayman Chemical Supplier No. 3022010 MG
A triterpene with diverse biological activities; inhibits ADP-, thrombin-, or epinephrine-induced aggregation of isolated rat platelets (IC50s = P. falciparum in vitro (IC50 = 4 µM); active against B. cereus, S. aureus, and S. pneumoniae (MICs = 39.1, 19.6, and 4.81 µM, respectively); cytotoxic to HepG2 cells (LC50 = 351 µM)