Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Acrylamidopropyl)trimethylammonium chloride solution

Catalog No. 44828150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-828-150ML 50 mL
44-828-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-828-150ML Supplier Sigma Aldrich Supplier No. 44828150ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Acrylamidopropyl)trimethylammonium chloride (APTAC) is a quaternary ammonium cationic monomer that is used as a temperature responsive copolymer.APTAC can be used in the formation of poly(APTAC) by graft polymerization, which can further be used as a flocculant.

Specifications

CAS 45021-77-0
Density 1.11 g/mL (at 25°C)
Molecular Formula C9H19ClN2O
Linear Formula H2C=CHCONH(CH2)3 N(CH3)3 Cl
Refractive Index n20/D 1.4848
Quantity 50 mL
MDL Number MFCD00274249
Synonym (Acrylamidopropyl)trimethylammonium chloride; Acryloylaminopropyltrimethylammonium chloride; Trimethyl(3-acrylamidopropyl)ammonium chloride
Molecular Weight (g/mol) 206.71
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.