Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Allyloxy-2-hydroxy-1-propanesulfonic acid sodium salt solution

Catalog No. 409421250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-942-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-942-1250ML Supplier Sigma Aldrich Supplier No. 409421250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Allyloxy-2-hydroxy-1-propanesulfonic acid sodium salt solution

Specifications

CAS 52556-42-0
Density 1.17 g/mL (at 25°C)
Molecular Formula C6H11NaO5S
Refractive Index n20/D 1.394
Linear Formula H2C=CHCH2OCH2CH(OH)CH2SO3Na
Quantity 250 mL
MDL Number MFCD00080396
Synonym 1-Propanesulfonic acid, 2-hydroxy-3-(2-propenyloxy)-, monosodium salt; Sodium 3-allyloxy-2-hydroxy-1-propanesulfonate
Molecular Weight (g/mol) 218.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.