Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Allyloxy)oxetane 97.0+%, TCI America™
SDP

Catalog No. A27945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27945G 5 g
A27941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27945G Supplier TCI America Supplier No. A27945G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6777-00-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD09908098
InChI Key NONFBYUIHZQHQT-UHFFFAOYSA-N
PubChem CID 81241
IUPAC Name 3-(prop-2-en-1-yloxy)oxetane
SMILES C=CCOC1COC1

Specifications

Color Colorless
Boiling Point 64°C
Quantity 5 g
UN Number 1993
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Allyloxy)oxetane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.