Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Allylsalicylaldehyde, 97%

Catalog No. p-7035748
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5347506 5 g
AAH5347514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5347506 Supplier Thermo Scientific Chemicals Supplier No. H5347506
Only null left
Add to Cart

CAS: 24019-66-7 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3-Allylsalicylaldehyde is a synthetic intermediate for the synthesis of 3-aminoflavones. Flavonoids have been studied for their antiproliferative activity and in vitro cytotoxicity. It is used in the preparation of oxazole derivatives and also imidazole scaffold-based 2-substituted benzofurans for application against human tumor cell lines. It is a reagent in the synthesis of antioxidant and antimicrobial novel chalcones.

Solubility
Soluble in chloroform.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24019-66-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00037364
InChI Key INLWEXRRMUMHKB-UHFFFAOYSA-N
Synonym 3-allyl-2-hydroxybenzaldehyde, 3-allylsalicylaldehyde, 3-allyl-2-hydroxy-benzaldehyde, benzaldehyde, 2-hydroxy-3-2-propenyl, salicylaldehyde, 3-allyl, 2-hydroxy-3-prop-2-en-1-yl benzaldehyde, acmc-209wev, tpc-sk001, 2-hydroxy-3-allylbenzaldehyde
PubChem CID 141062
IUPAC Name 2-hydroxy-3-prop-2-enylbenzaldehyde
SMILES C=CCC1=CC=CC(=C1O)C=O

Specifications

Density 1.099
Flash Point 107°C (225°F)
Refractive Index 1.5645
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 3-Allylsalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.