Learn More
Alkali Scientific Alkali Scientific™ 3″ Aluminum Box for Storage of Samples or Tubes with 4 Drain Holes, 81 Cell Divider
SDP
Supplier:  Alkali Scientific FB3A81
Encompass_Preferred
Aluminum Cryovial boxes come with or without cardboard dividers and fit into standard freezer racks. These boxes conform to international standard dimensions and hold 2 to 5 milliliter cryo vials or centrifuge tubes. They can be used as cryo boxes as well as regular storage boxes. 3" Stainless steel freezer storage boxes with multiple options for cell divider included. Fits into standard freezer racks. Can be used as cryo boxes as well as regular storage boxes. Has drain holes.  Product features 3" Aluminum box with drain holes Holds 2 to 5 milliliter cryo vials or centrifuge tubes Can be used as cryo boxes as well as regular storage boxes Fits into standard freezer racks Measures 5-1/4" length by 5-1/4" width by 3" height
Catalog No. 50-302-7722