Learn More
Cayman Chemical 3-Allylphenol, Purity: ≥98%, CAS: 1446-24-8, 10 mg
SDP
Catalog No. 502912062
Encompass_Preferred
An inhibitor of dopamine β-hydroxylase (Ki = 12 mM)
Catalog No. 50-291-2062 Supplier Cayman Chemical Supplier No. 3649210 MG
An inhibitor of dopamine β-hydroxylase (Ki = 12 mM)