Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1,2,4-dithiazole-5-thione, 98%, Thermo Scientific™

Catalog No. AAA1065803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1065803 1 g
AAA1065806 5 g
AAA1065814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1065803 Supplier Thermo Scientific Chemicals Supplier No. A1065803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6846-35-1
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.232
MDL Number MFCD00051660
InChI Key YWZHEXZIISFIDA-UHFFFAOYSA-N
Synonym xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol
PubChem CID 2735329
IUPAC Name 5-amino-1,2,4-dithiazole-3-thione
SMILES C1(=NC(=S)SS1)N

Specifications

Melting Point 201°C to 203°C
Quantity 1 g
Formula Weight 150.26
Percent Purity 98%
Chemical Name or Material 3-Amino-1,2,4-dithiazole-5-thione
Hazard Category Warning
Hazard Statement GHS H Statement
H373-H315-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber JO6500000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.