Learn More
Medchemexpress LLC 3-Amino-1,2,4-triazine | 1120-99-6 | 96.09 | 25 G
SDP
Catalog No. 5000448140
Encompass_Preferred
3-Amino-1,2,4-triazine is an inhibitor of NO synthase and inhibits nitrite secretion. It is intended for research use only.
  • Inhibits NO synthase activity in cytosolic extracts of activated cells
  • Inhibits nitrite secretion by activated macrophages
  • Inhibits parasite killing by activated macrophages
Catalog No. 50-004-48140 Supplier Medchemexpress LLC Supplier No. 25GHYW004843
3-Amino-1,2,4-triazine is an inhibitor of NO synthase and inhibits nitrite secretion. It is intended for research use only.
  • Inhibits NO synthase activity in cytosolic extracts of activated cells
  • Inhibits nitrite secretion by activated macrophages
  • Inhibits parasite killing by activated macrophages