Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-1,2,4-triazine | 1120-99-6 | 96.09 | 5 G
SDP

Catalog No. 5000448997
Encompass_Preferred

3-Amino-1,2,4-triazine is an inhibitor of NO synthase and also inhibits nitrite secretion.

  • Inhibits NO synthase activity in cytosolic extracts of activated cells
  • Inhibits nitrite secretion and parasite killing by activated macrophages
  • Appears as a solid
  • Color ranges from brown to black
  • Soluble in DMSO at 100 mg/mL
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-48997 Supplier Medchemexpress LLC Supplier No. HYW0048435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Amino-1,2,4-triazine is an inhibitor of NO synthase and also inhibits nitrite secretion.

  • Inhibits NO synthase activity in cytosolic extracts of activated cells
  • Inhibits nitrite secretion and parasite killing by activated macrophages
  • Appears as a solid
  • Color ranges from brown to black
  • Soluble in DMSO at 100 mg/mL
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.