Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1,2,4-triazole, 96.1%, MP Biomedicals™

Catalog No. ICN10064710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10064710 10 g
ICN10064725 25 g
ICN10064780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10064710 Supplier MP Biomedicals, Inc Supplier No. 0210064710
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61-82-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00005230,MFCD00053362
InChI Key KLSJWNVTNUYHDU-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
PubChem CID 1639
ChEBI CHEBI:40036
IUPAC Name 1H-1,2,4-triazol-5-amine
SMILES NC1=NC=NN1

Specifications

Color White
Density ≥0.6 to 0.85g/cm3 at 20°C
Assay Percent Range 0.961
Quantity 10 g
Formula Weight 84.08
Physical Form Powder
Chemical Name or Material 3-Amino-1, 2, 4-triazole
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.