Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-3-Amino-1,2-propanediol

Catalog No. A7600125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A7600125G 25 g
A76001100G 100 g
A760015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A7600125G Supplier Sigma Aldrich Supplier No. A7600125G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • (+/-)-3-amino-1,2-propanediol is used as a reactant in the synthesis of lipid-like delivery molecules(lipidoids) for RNA interference (rnai) therapeutics
  • It is also used in the synthesis of the bioactive template to prepare cationic α-helical polypeptides and various cationic polymers for gene delivery

Specifications

CAS 616-30-8
Density 1.175 g/mL (at 25°C (literature))
Boiling Point 264°C to 265°C (739 mmHg)
Molecular Formula C3H9NO2
Refractive Index n20/D 1.492 (literature)
Linear Formula NH2CH2CH(OH)CH2OH
MDL Number MFCD00008140
Quantity 25 g
Molecular Weight (g/mol) 91.11
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.