Learn More
Medchemexpress LLC (±)-3-Amino-1,2-propanediol | 616-30-8 | 91.11 | 100 G
SDP
Catalog No. 5000447093
Encompass_Preferred
(±)-3-Amino-1,2-propanediol, also known as 1-aminoglycerol, is a chiral raw material used in the synthesis of chiral vanadium (V) Schiff base complexes.
  • Used as a chiral raw material
  • Suitable for synthesis of chiral vanadium (V) Schiff base complexes
  • Hygroscopic
  • Synthetic origin, free from animal byproducts
  • Store at 4°C, protected from light
  • Stock solution stable at -80°C for 6 months, or -20°C for 1 month (protected from light)
  • Appears as a solid below 55°C, liquid above 57°C
Catalog No. 50-004-47093 Supplier Medchemexpress LLC Supplier No. HYW002040100G
(±)-3-Amino-1,2-propanediol, also known as 1-aminoglycerol, is a chiral raw material used in the synthesis of chiral vanadium (V) Schiff base complexes.
  • Used as a chiral raw material
  • Suitable for synthesis of chiral vanadium (V) Schiff base complexes
  • Hygroscopic
  • Synthetic origin, free from animal byproducts
  • Store at 4°C, protected from light
  • Stock solution stable at -80°C for 6 months, or -20°C for 1 month (protected from light)
  • Appears as a solid below 55°C, liquid above 57°C