Learn More
Medchemexpress LLC (±)-3-Amino-1,2-propanediol | 616-30-8 | 91.11 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW002040
Encompass_Preferred
(±)-3-Amino-1,2-propanediol, also known as 1-Aminoglycerol, is a chiral raw material.
  • Used for the synthesis of chiral vanadium (V) Schiff base complexes.
  • Intended for research use only.
  • Not for sale to patients.
Catalog No. 50-004-49942