Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-benzylpiperidine 98.0+%, TCI America™
SDP

Catalog No. A29171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29171G 1 g
A29175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29171G Supplier TCI America Supplier No. A29171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60407-35-4
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD04038444
InChI Key HARWNWOLWMTQCC-UHFFFAOYSA-N
PubChem CID 421694
IUPAC Name 1-benzylpiperidin-3-amine
SMILES C1CC(CN(C1)CC2=CC=CC=C2)N

Specifications

Color White-Yellow
Boiling Point 82°C
Quantity 1 g
Formula Weight 190.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Amino-1-benzylpiperidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.