Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-propanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019562 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9562 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9562 Supplier MilliporeSigma Supelco Supplier No. PHR1339500MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 770°F
CAS 156-87-6
Vapor Pressure 2.1 mmHg (20°C)
Vapor Density 2.59 (vs. air)
Density 0.982 g/mL (at 20°C)
Flash Point 100°C - Pensky-Martens°Closed°Cup
Physical Form Neat
Linear Formula HO(CH2)3 NH2
MDL Number MFCD00008223
Melting Point 10°C to 12°C
Boiling Point 184°C to 187°C
Synonym 3-Amino-1-propanol; 3-Aminopropyl alcohol
Type Certified Reference Material
Quantity 500 mg
Formula Weight 75.11
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.