Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,4,6-triiodobenzoic acid, 99+%, Thermo Scientific™

Catalog No. AC277650250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC277650250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC277650250 Supplier Thermo Scientific Chemicals Supplier No. 277650250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3119-15-1
Molecular Formula C7H3I3NNaO2
Molecular Weight (g/mol) 536.81
MDL Number MFCD00007681
InChI Key CTXRMWPLTDAHOR-UHFFFAOYSA-M
Synonym unii-99h77d8mtd, 3-amino-2,4,6-triiodo-benzoic acid, acido 3-amino-2,4,6-triiodobenzoico italian, benzoic acid, 3-amino-2,4,6-triiodo, 3-amino-2,4,6-triiodo-benzoicacid, acido 3-amino-2,4,6-triiodobenzoico, pubchem4987, acmc-1co9w, 4-14-00-01118 beilstein handbook reference, 2,4,6-triiodo-3-aminobenzoic acid
PubChem CID 18387
IUPAC Name sodium 3-amino-2,4,6-triiodobenzoate
SMILES [Na+].NC1=C(I)C=C(I)C(C([O-])=O)=C1I

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 196°C to 199°C
Color Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula H2NC6H(I)3CO2H
Quantity 25 g
Beilstein 14, 414
Formula Weight 514.83
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material 3-Amino-2, 4, 6-triiodobenzoic acid, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.