Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,5-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. A24445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24445G Supplier TCI America Supplier No. A24445G

Chemical Identifiers

CAS 78607-32-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00204157
InChI Key OLZPJUVEGSNIJL-UHFFFAOYSA-N
Synonym 3-amino-2,5-dichloropyridine, 2,5-dichloro-3-aminopyridine, 2,5-dichloro-pyridin-3-ylamine, 3-pyridinamine, 2,5-dichloro, 2,5-dichloro-3-pyridylamine, 2,5-dichloropyridine-3-amine, pubchem5507, acmc-209per, ksc495k8j
PubChem CID 2799999
IUPAC Name 2,5-dichloropyridin-3-amine
SMILES C1=C(C(=NC=C1Cl)Cl)N

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,5-dichloropyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.