Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,6-difluorobenzoic acid, 95%, Thermo Scientific™

Catalog No. AAH6664503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6664503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6664503 Supplier Thermo Scientific Chemicals Supplier No. H6664503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83141-11-1
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD07368833
InChI Key ORSUTLAFOCNIDY-UHFFFAOYSA-N
PubChem CID 18405352
IUPAC Name 3-amino-2,6-difluorobenzoic acid
SMILES C1=CC(=C(C(=C1N)F)C(=O)O)F

Specifications

Quantity 1 g
Beilstein 13330504
Synonym 3-amino-2,6-difluorobenzoicacid, pubchem8200, 3-amino-2,6-difluoro-benzoic acid, buttpark 120\01-13, benzoic acid,3-amino-2,6-difluoro, benzoic acid, 3-amino-2,6-difluoro
Formula Weight 173.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Amino-2,6-difluorobenzoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.