Learn More
Cayman Chemical 3-Amino-2,6-Piperidinedione, Purity: ≥95%, CAS: 2353-44-8, 50 mg
SDP
Catalog No. 502978925
Encompass_Preferred
An active metabolite of (±)-thalidomide; increases hexobarbitone-induced sleeping time in rats at 400 mg/kg
Catalog No. 50-297-8925 Supplier Cayman Chemical Supplier No. 3169850 MG
An active metabolite of (±)-thalidomide; increases hexobarbitone-induced sleeping time in rats at 400 mg/kg