Learn More
Medchemexpress LLC 3-Amino-2,6-piperidinedione | 2353-44-8 | 100.0% | 128.13 | 10 G
SDP
Catalog No. 5000405064
Encompass_Preferred
3-Amino-2,6-piperidinedione is a synthetic intermediate used in the design of antitumor compounds. This product is intended for research use only and has not been fully validated for medical applications.
  • Synthetic intermediate for antitumor compound design
  • Stable under recommended storage conditions
  • Should be stored at 4°C, protected from light
  • For research use only
Catalog No. 50-004-05064 Supplier Medchemexpress LLC Supplier No. HY3459010G
3-Amino-2,6-piperidinedione is a synthetic intermediate used in the design of antitumor compounds. This product is intended for research use only and has not been fully validated for medical applications.
  • Synthetic intermediate for antitumor compound design
  • Stable under recommended storage conditions
  • Should be stored at 4°C, protected from light
  • For research use only