Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-2,6-piperidinedione | 2353-44-8 | 99.96% | 128.13 | 500 MG
SDP

Catalog No. 5000403771
Encompass_Preferred

3-Amino-2,6-piperidinedione is a synthetic intermediate utilized in the design of antitumor compounds. This product is for research use only and is not sold to patients.

  • Synthetic intermediate.
  • Used in the design of antitumor compounds.
  • Purity: 99.96%.
  • Available in multiple pack sizes (e.g., 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g).
  • Appearance: Solid, off-white to light blue.
  • Shipping: Room temperature (continental US).
  • Storage: 4°C, protected from light.
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protected from light).
  • For research use only.
  • Cited in scientific literature for its use in synthesizing antitumor agents.

Catalog No. 50-004-03771 Supplier Medchemexpress LLC Supplier No. HY34590500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-2,6-piperidinedione is a synthetic intermediate utilized in the design of antitumor compounds. This product is for research use only and is not sold to patients.

  • Synthetic intermediate.
  • Used in the design of antitumor compounds.
  • Purity: 99.96%.
  • Available in multiple pack sizes (e.g., 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g).
  • Appearance: Solid, off-white to light blue.
  • Shipping: Room temperature (continental US).
  • Storage: 4°C, protected from light.
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protected from light).
  • For research use only.
  • Cited in scientific literature for its use in synthesizing antitumor agents.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.