Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-2-hydroxypropanoic acid | 565-71-9 | 98.0% | 105.09 | 25 G
SDP

Catalog No. 5000451383
Encompass_Preferred

3-Amino-2-hydroxypropanoic acid is a serine derivative used in research. It is recognized as a beneficial ergogenic dietary substance due to its influence on anabolic hormones, fuel supply during exercise, mental performance, and prevention of muscle damage.

  • For research use only
  • Appearance: Solid, white to off-white
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in H2O: 10 mg/mL (95.16 mM; requires ultrasonic)

Catalog No. 50-004-51383 Supplier Medchemexpress LLC Supplier No. 25GHYW012966
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Amino-2-hydroxypropanoic acid is a serine derivative used in research. It is recognized as a beneficial ergogenic dietary substance due to its influence on anabolic hormones, fuel supply during exercise, mental performance, and prevention of muscle damage.

  • For research use only
  • Appearance: Solid, white to off-white
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in H2O: 10 mg/mL (95.16 mM; requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.