Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-methylacrylaldehyde, 97%, Thermo Scientific™

Catalog No. RH01431EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RH01431EA 10 g Amber Glass Bottle
RH01431DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RH01431EA Supplier Thermo Scientific Chemicals Supplier No. RH01431EA
Add to Cart

Chemical Identifiers

CAS 30989-81-2
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD01566866
InChI Key OEZAAXHZEMTBOV-DUXPYHPUSA-N
Synonym 3-amino-2-methylacrylaldehyde, 3-amino-2-methylacrolein, 2e-3-amino-2-methylprop-2-enal, amino-alpha-methylacrolein, e-3-amino-2-methylprop-2-enal, 3-amino-2-methyl acrylaldehyde, 2-propenal, 3-amino-2-methyl-, e
PubChem CID 637539
IUPAC Name (E)-3-amino-2-methylprop-2-enal
SMILES CC(=CN)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula NH2CH=C(CH3)CHO
Quantity 10 g
Formula Weight 85.11
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methylacrylaldehyde
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.