Learn More
Medchemexpress LLC 3-Amino-2-naphthoic acid | 5959-52-4 | 187.20 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0051481G
Encompass_Preferred
3-Amino-2-naphthoic acid is a derivative of naphthalene that can be used to synthesize other compounds. It is commonly utilized as a base material for synthesizing various dyes and pigments. This product has also been developed as a "turn-on" fluorescence probe for the specific detection of the cyanate anion (CNO-).
  • Derivative of naphthalene.
  • Used to synthesize other compounds.
  • Functions as a base material for various dyes and pigments.
  • Developed as a "turn-on" fluorescence probe.
  • Specific detection of the cyanate anion (CNO-).
Catalog No. 50-004-45494