Learn More
Medchemexpress LLC 3-Amino-2-naphthoic acid | 5959-52-4 | MFCD00004115 | 187.20 | 5 G
SDP
Catalog No. 5000446045
Encompass_Preferred
3-Amino-2-naphthoic acid is a naphthalene derivative that can be utilized in the synthesis of other compounds. It is commonly employed as a base material for the creation of various dyes and pigments. Additionally, 3-Amino-2-naphthoic acid has been developed as a "turn-on" fluorescence probe for the specific detection of the cyanate anion (CNO-).
  • Utilized in the synthesis of other compounds
  • Commonly employed as a base material for the creation of various dyes and pigments
  • Developed as a "turn-on" fluorescence probe for the specific detection of the cyanate anion (CNO-)
Catalog No. 50-004-46045 Supplier Medchemexpress LLC Supplier No. HYW0051485G
3-Amino-2-naphthoic acid is a naphthalene derivative that can be utilized in the synthesis of other compounds. It is commonly employed as a base material for the creation of various dyes and pigments. Additionally, 3-Amino-2-naphthoic acid has been developed as a "turn-on" fluorescence probe for the specific detection of the cyanate anion (CNO-).
  • Utilized in the synthesis of other compounds
  • Commonly employed as a base material for the creation of various dyes and pigments
  • Developed as a "turn-on" fluorescence probe for the specific detection of the cyanate anion (CNO-)