Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone | 80-65-9 | 98.0% | 102.09 | 10 MG
SDP
Catalog No. 5000454089
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone used as an indicator for furazolidone residues in vivo. It is orally active and inhibits monoamine oxidase (MAO), increasing monoamine neurotransmitter levels in the brain and lowering blood pressure.
  • Used as an indicator for furazolidone residues
  • Orally active
  • Inhibits monoamine oxidase (MAO)
  • Increases levels of monoamine neurotransmitters
  • Lowers blood pressure in renal hypertensive rats
Catalog No. 50-004-54089 Supplier Medchemexpress LLC Supplier No. HYW01298210MG
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone used as an indicator for furazolidone residues in vivo. It is orally active and inhibits monoamine oxidase (MAO), increasing monoamine neurotransmitter levels in the brain and lowering blood pressure.
  • Used as an indicator for furazolidone residues
  • Orally active
  • Inhibits monoamine oxidase (MAO)
  • Increases levels of monoamine neurotransmitters
  • Lowers blood pressure in renal hypertensive rats