Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone | 80-65-9 | 98.0% | 102.09 | 5 MG
SDP
Catalog No. 5000451454
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is used as an indicator for furazolidone residues in vivo and is orally active. It inhibits monoamine oxidase (MAO) via its metabolite in rat brain and liver. It also increases levels of monoamine neurotransmitters (such as norepinephrine and dopamine) in the brain and lowers blood pressure in renal hypertensive rats.
  • Metabolite of furazolidone.
  • Always detected as an indicator of furazolidone residues in vivo.
  • Orally active.
  • Inhibits monoamine oxidase (MAO) via its metabolite in rat brain and liver.
  • Increases levels of monoamine neurotransmitters (such as norepinephrine and dopamine) in the brain.
  • Lowers blood pressure in renal hypertensive rats.
Catalog No. 50-004-51454 Supplier Medchemexpress LLC Supplier No. HYW0129825MG
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is used as an indicator for furazolidone residues in vivo and is orally active. It inhibits monoamine oxidase (MAO) via its metabolite in rat brain and liver. It also increases levels of monoamine neurotransmitters (such as norepinephrine and dopamine) in the brain and lowers blood pressure in renal hypertensive rats.
  • Metabolite of furazolidone.
  • Always detected as an indicator of furazolidone residues in vivo.
  • Orally active.
  • Inhibits monoamine oxidase (MAO) via its metabolite in rat brain and liver.
  • Increases levels of monoamine neurotransmitters (such as norepinephrine and dopamine) in the brain.
  • Lowers blood pressure in renal hypertensive rats.