Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone | 80-65-9 | MFCD00020871 | 98.0% | 102.09 | 1 ML
SDP
Catalog No. 5000367008
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is consistently detected as an indicator of furazolidone residues in vivo. This compound is orally active.
  • It is a metabolite of furazolidone.
  • It is an indicator of furazolidone residues in vivo.
  • The compound is orally active.
  • It inhibits monoamine oxidase (MAO) via its metabolite in rats' brain and liver.
  • It increases levels of monoamine neurotransmitters like norepinephrine and dopamine in the brain.
  • It lowers blood pressure in renal hypertensive rats.
Catalog No. 50-003-67008 Supplier Medchemexpress LLC Supplier No. HYW01298210MM/1ML
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is consistently detected as an indicator of furazolidone residues in vivo. This compound is orally active.
  • It is a metabolite of furazolidone.
  • It is an indicator of furazolidone residues in vivo.
  • The compound is orally active.
  • It inhibits monoamine oxidase (MAO) via its metabolite in rats' brain and liver.
  • It increases levels of monoamine neurotransmitters like norepinephrine and dopamine in the brain.
  • It lowers blood pressure in renal hypertensive rats.