Learn More
Medchemexpress LLC 3-amino-2-oxazolidinone (AOZ) | 80-65-9 | 98.0% | 102.09 | 25 MG
SDP
Catalog No. 5000452665
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is used as an indicator for furazolidone residues in vivo and is orally active. Research indicates it inhibits monoamine oxidase (MAO) through its metabolite in rat brain and liver, increasing monoamine neurotransmitter levels like norepinephrine and dopamine, and lowering blood pressure in renal hypertensive rats. It has been observed to reduce blood pressure in renal hypertensive rat models by 60% at a dosage of 30 mg/kg administered orally once daily for 10 days. This product is for research use only and not sold to patients.
  • Metabolite of furazolidone
  • Used as an indicator for furazolidone residues in vivo
  • Orally active
  • Inhibits monoamine oxidase (MAO)
  • Increases monoamine neurotransmitter levels
  • Lowers blood pressure in renal hypertensive rats
  • For research use only
Catalog No. 50-004-52665 Supplier Medchemexpress LLC Supplier No. 25MGHYW012982S
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone. It is used as an indicator for furazolidone residues in vivo and is orally active. Research indicates it inhibits monoamine oxidase (MAO) through its metabolite in rat brain and liver, increasing monoamine neurotransmitter levels like norepinephrine and dopamine, and lowering blood pressure in renal hypertensive rats. It has been observed to reduce blood pressure in renal hypertensive rat models by 60% at a dosage of 30 mg/kg administered orally once daily for 10 days. This product is for research use only and not sold to patients.
  • Metabolite of furazolidone
  • Used as an indicator for furazolidone residues in vivo
  • Orally active
  • Inhibits monoamine oxidase (MAO)
  • Increases monoamine neurotransmitter levels
  • Lowers blood pressure in renal hypertensive rats
  • For research use only