Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone (AOZ) | 80-65-9 | 98.0% | 102.09 | 50 MG
SDP
Catalog No. 5000451389
Encompass_Preferred
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone and is used as an indicator for furazolidone residues in vivo. This orally active product inhibits monoamine oxidase (MAO) through its metabolite in rat brain and liver, increasing levels of monoamine neurotransmitters like norepinephrine and dopamine, and lowering blood pressure in renal hypertensive rats.
  • Used as an indicator for furazolidone residues
  • Orally active
  • Increases levels of monoamine neurotransmitters
  • Lowers blood pressure in renal hypertensive rats
Catalog No. 50-004-51389 Supplier Medchemexpress LLC Supplier No. HYW012982S50MG
3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone and is used as an indicator for furazolidone residues in vivo. This orally active product inhibits monoamine oxidase (MAO) through its metabolite in rat brain and liver, increasing levels of monoamine neurotransmitters like norepinephrine and dopamine, and lowering blood pressure in renal hypertensive rats.
  • Used as an indicator for furazolidone residues
  • Orally active
  • Increases levels of monoamine neurotransmitters
  • Lowers blood pressure in renal hypertensive rats