Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone-d4 | 1188331-23-8 | 99.9% | 106.12 | 5 MG
SDP
Catalog No. 5000451993
Encompass_Preferred
3-Amino-2-oxazolidinone-d4 (AOZ-d4) is a deuterium labeled 3-Amino-2-oxazolidinone, which is the metabolite of Furazolidone. 3-Amino-2-oxazolidinone is always detected as an indicator of furazolidone residues in vivo and is orally active. It can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Orally active
Catalog No. 50-004-51993 Supplier Medchemexpress LLC Supplier No. HYW012982S5MG
3-Amino-2-oxazolidinone-d4 (AOZ-d4) is a deuterium labeled 3-Amino-2-oxazolidinone, which is the metabolite of Furazolidone. 3-Amino-2-oxazolidinone is always detected as an indicator of furazolidone residues in vivo and is orally active. It can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Orally active