Learn More
Medchemexpress LLC 3-Amino-2-oxazolidinone-d4 (AOZ-d4) | 1188331-23-8 | 99.9% | 106.12 | 1 MG
SDP
Catalog No. 5000451455
Encompass_Preferred
3-Amino-2-oxazolidinone-d4 (AOZ-d4) is a deuterium labeled version of 3-Amino-2-oxazolidinone (HY-W012982). 3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone (HY-B1336) and serves as an indicator of furazolidone residues in vivo. It is orally active and can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. The product appears as a white to off-white solid. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51455 Supplier Medchemexpress LLC Supplier No. HYW012982S1MG
3-Amino-2-oxazolidinone-d4 (AOZ-d4) is a deuterium labeled version of 3-Amino-2-oxazolidinone (HY-W012982). 3-Amino-2-oxazolidinone (AOZ) is a metabolite of Furazolidone (HY-B1336) and serves as an indicator of furazolidone residues in vivo. It is orally active and can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. The product appears as a white to off-white solid. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.