Learn More
Medchemexpress LLC 3-Amino-2-piperidinone | 1892-22-4 | 99.87% | 114.15 | 1 ML
SDP
Catalog No. 5000402122
Encompass_Preferred
3-Amino-2-piperidinone is a cyclic ornithine analogue that can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, an inhibitor of Fas-induced Apoptosis.
  • Functions as a cyclic ornithine analogue.
  • Can be used in the synthesis of N-(2-oxopiperidin-3-yl)dodecanamide.
  • Inhibits Fas-induced Apoptosis.
Catalog No. 50-004-02122 Supplier Medchemexpress LLC Supplier No. HY2108810MM/1ML
3-Amino-2-piperidinone is a cyclic ornithine analogue that can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, an inhibitor of Fas-induced Apoptosis.
  • Functions as a cyclic ornithine analogue.
  • Can be used in the synthesis of N-(2-oxopiperidin-3-yl)dodecanamide.
  • Inhibits Fas-induced Apoptosis.