Learn More
Medchemexpress LLC 3-Amino-2-piperidinone | 1892-22-4 | 99.9% | 114.15 | 25 G
SDP
Catalog No. 5000402842
Encompass_Preferred
3-Amino-2-piperidinone is a cyclic ornithine analogue for research and analytical applications. It can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, which inhibits Fas-induced Apoptosis. It is classified as a Drug Intermediate and Reference Standard.
  • Cyclic ornithine analogue
  • Used as a drug intermediate
  • Intended for research and analytical applications
  • Inhibits Fas-induced Apoptosis
Catalog No. 50-004-02842 Supplier Medchemexpress LLC Supplier No. 25GHY21088
3-Amino-2-piperidinone is a cyclic ornithine analogue for research and analytical applications. It can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, which inhibits Fas-induced Apoptosis. It is classified as a Drug Intermediate and Reference Standard.
  • Cyclic ornithine analogue
  • Used as a drug intermediate
  • Intended for research and analytical applications
  • Inhibits Fas-induced Apoptosis