Learn More
Medchemexpress LLC 3-Amino-2-piperidinone | 1892-22-4 | 99.93% | 114.15 | 500 MG
SDP
Catalog No. 5000401028
Encompass_Preferred
3-amino-2-piperidinone is a cyclic ornithine analogue that can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, a compound known to inhibit Fas-induced Apoptosis. This product is intended for research use only and is available in various solid and solution quantities.
  • Cyclic ornithine analogue.
  • Precursor for synthesizing N-(2-oxopiperidin-3-yl)dodecanamide.
  • Available in various quantities.
  • Available in solution and solid forms.
  • For research use only.
Catalog No. 50-004-01028 Supplier Medchemexpress LLC Supplier No. HY21088500MG
3-amino-2-piperidinone is a cyclic ornithine analogue that can be used to synthesize N-(2-oxopiperidin-3-yl)dodecanamide, a compound known to inhibit Fas-induced Apoptosis. This product is intended for research use only and is available in various solid and solution quantities.
  • Cyclic ornithine analogue.
  • Precursor for synthesizing N-(2-oxopiperidin-3-yl)dodecanamide.
  • Available in various quantities.
  • Available in solution and solid forms.
  • For research use only.