Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(3-pyridyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. A25371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25371G Supplier TCI America Supplier No. A25371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62247-21-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD01549598
InChI Key QOTCEJINJFHMLO-UHFFFAOYNA-N
PubChem CID 3606547
IUPAC Name 3-amino-3-(pyridin-3-yl)propanoic acid
SMILES NC(CC(O)=O)C1=CN=CC=C1

Specifications

Melting Point 207°C
Color White
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(3-pyridyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.