Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(4-fluorophenyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. A24301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24301G Supplier TCI America Supplier No. A24301G

Chemical Identifiers

CAS 325-89-3
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00181811
InChI Key CPGFMWPQXUXQRX-UHFFFAOYSA-N
Synonym 3-amino-3-4-fluorophenyl propanoic acid, 3-amino-3-4-fluorophenyl propionic acid, 3-amino-3-4-fluoro-phenyl-propionic acid, 3-p-fluorophenyl-dl-beta-alanine, r-3-p-fluorophenyl-beta-alanine, 3-4-fluorophenyl-beta-alanine, dl-3-amino-3-4-fluoro-phenyl-propionic acid, pubchem13896, pubchem17300, acmc-20a8bw
PubChem CID 579885
IUPAC Name 3-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)N)F

Specifications

Melting Point 216°C
Color White
Quantity 1 g
Formula Weight 183.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(4-fluorophenyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.