Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 3-Amino-3-Deoxydigoxigenin Hemisuccinamide, Succinimidyl Ester

Catalog No. A2952
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2952 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2952 Supplier Invitrogen™ Supplier No. A2952
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

3-amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester

The amine-reactive version of digoxigenin (3-amino-3-deoxygigoxigenin hemisuccinamide, SE) can be used to attach this important hapten to biomolecules of interest for subsequent detection with an anti-digoxigenin antibody.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type 3-Amino-3-Deoxydigoxigenin Hemisuccinamide Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Digoxigenin
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.