Learn More
Cayman Chemical 3'-Amino-3'-deoxyguanosine-5'-O-triphosphate (sodium salt), Purity: ≥95%, 2.6 mg
SDP
Catalog No. 502918999
Encompass_Preferred
An amine-modified nucleotide; terminates RNA strand synthesis catalyzed by RNA polymerase in E. coli; has been used in the synthesis of a non-hydrolyzable derivative of pppGpp
Catalog No. 50-291-8999 Supplier Cayman Chemical Supplier No. 385432.6 MG
An amine-modified nucleotide; terminates RNA strand synthesis catalyzed by RNA polymerase in E. coli; has been used in the synthesis of a non-hydrolyzable derivative of pppGpp