Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-phenyl-1-propanol 97.0+%, TCI America™
SDP

Catalog No. A25345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25345G 5 g
A25341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25345G Supplier TCI America Supplier No. A25345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14593-04-5
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00801124,MFCD01311768,MFCD01311791
InChI Key SEQXIQNPMQTBGN-UHFFFAOYNA-N
Synonym 3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0
PubChem CID 203500
IUPAC Name 3-amino-3-phenylpropan-1-ol
SMILES NC(CCO)C1=CC=CC=C1

Specifications

Melting Point 75°C
Color White
Boiling Point 134°C
Quantity 5 g
UN Number 3259
Formula Weight 151.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-phenyl-1-propanol
RTECSNumber UA6903000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.