Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-amino-4-(3-hexylphenylamino)-4-oxobutyl phosphonic acid | 909725-62-8 | 98.2% | 456.39 g/mol | C18H28F3N2O6P | 1 ML
SDP

Catalog No. 5000212466
Encompass_Preferred

W146 TFA is the trifluoroacetic acid salt of W146, a selective antagonist of the sphingosine-1-phosphate receptor 1 (S1PR1). It is supplied as a ready-to-use 10 mM solution in DMSO and as solid material for research applications, and is used in in vitro pharmacology and signaling studies to probe S1PR1-mediated pathways.

  • Selective antagonist of sphingosine-1-phosphate receptor 1 (S1PR1).
  • Reported EC50 of 398 nM.
  • Supplied as 10 mM solution in DMSO (1 mL) and in multiple solid masses.
  • Chemical identifiers: CAS 909725-62-8, formula C18H28F3N2O6P, MW 456.39 g/mol.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-12466 Supplier Medchemexpress LLC Supplier No. HY101395A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

W146 TFA is the trifluoroacetic acid salt of W146, a selective antagonist of the sphingosine-1-phosphate receptor 1 (S1PR1). It is supplied as a ready-to-use 10 mM solution in DMSO and as solid material for research applications, and is used in in vitro pharmacology and signaling studies to probe S1PR1-mediated pathways.

  • Selective antagonist of sphingosine-1-phosphate receptor 1 (S1PR1).
  • Reported EC50 of 398 nM.
  • Supplied as 10 mM solution in DMSO (1 mL) and in multiple solid masses.
  • Chemical identifiers: CAS 909725-62-8, formula C18H28F3N2O6P, MW 456.39 g/mol.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.