Learn More
Medchemexpress LLC 3-amino-4-{(3S)-3-[(2-ethoxyethoxy)methyl]piperidin-1-yl}thieno[2,3-b]pyridine-2-carboxamide | 2871872-79-4 | 99.9% | 378.49 | C18H26N4O3S | 5 MG
SDP
Catalog No. 5000221710
Encompass_Preferred
DS96432529 is a small-molecule cyclin-dependent kinase 8 (CDK8) inhibitor reported to have potent, orally active bone anabolic effects. It is supplied for research use and is characterized by a molecular formula of C18H26N4O3S, a molecular weight of 378.49 g/mol, and a CAS number of 2871872-79-4. Analytical documentation and a certificate of analysis are available.
  • Potent CDK8 inhibition with bone anabolic activity.
  • Orally active pharmacological profile in reported studies.
  • High purity (99.9%) suitable for research applications.
  • Available in small research pack sizes, including 5 mg.
  • Characterized by formula C18H26N4O3S and molecular weight 378.49 g/mol.
  • Analytical documentation available (COA, H NMR, LC-MS).
Catalog No. 50-002-21710 Supplier Medchemexpress LLC Supplier No. HY1451215MG
DS96432529 is a small-molecule cyclin-dependent kinase 8 (CDK8) inhibitor reported to have potent, orally active bone anabolic effects. It is supplied for research use and is characterized by a molecular formula of C18H26N4O3S, a molecular weight of 378.49 g/mol, and a CAS number of 2871872-79-4. Analytical documentation and a certificate of analysis are available.
  • Potent CDK8 inhibition with bone anabolic activity.
  • Orally active pharmacological profile in reported studies.
  • High purity (99.9%) suitable for research applications.
  • Available in small research pack sizes, including 5 mg.
  • Characterized by formula C18H26N4O3S and molecular weight 378.49 g/mol.
  • Analytical documentation available (COA, H NMR, LC-MS).