Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-bromobenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. A14835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14835G Supplier TCI America Supplier No. A14835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 454-79-5
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
MDL Number MFCD00007637
InChI Key PZDVFXUBTKPFSG-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl aniline, 3-amino-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl benzenamine, 2-bromo-5-trifluoromethylphenylamine, 2-amino-1-bromo-4-trifluoromethylbenzene, 2-amino-4-trifluoromethylbromobenzene, 6-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, 3-amino-4-bromo trifluoromethyl benzene, benzenamine, 2-bromo-5-trifluoromethyl, 4-bromo-3-aminobenzotrifluoride
PubChem CID 67989
IUPAC Name 2-bromo-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)Br

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 240.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Amino-4-bromobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.