Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-chlorobenzonitrile 97.0+%, TCI America™
SDP

Catalog No. A21055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21055G Supplier TCI America Supplier No. A21055G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53312-79-1
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00017089
InChI Key KRBRYJLBILJHAS-UHFFFAOYSA-N
Synonym 2-Chloro-5-cyanoaniline
PubChem CID 104464
IUPAC Name 3-amino-4-chlorobenzonitrile
SMILES NC1=CC(=CC=C1Cl)C#N

Specifications

Melting Point 90°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 152.58
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3-Amino-4-chlorobenzonitrile
EINECSNumber (3)-1817
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.